Lab Sinks, Self Rimming, Three Bowl, V-3 Series, Eagle MHC
Supplier: Eagle Group
Certificates
About this item
Heavy gauge type 304 series stainless steel coved bowls with large radius.
- Three compartment
- 1½" Ø basket drain in center of bowl
Sink can be mounted on top or bottom of countertop. Features ¾" hole for DI faucet, 1³/₈" hole for mixing faucet and 1¼" hole for eye/drench hose.
Frequently Bought Together