Lab Sinks, Self Rimming, Two Bowl, V-3 Series, Eagle MHC
Supplier: Eagle Group
Certificates
About this item
Heavy gauge type 304 series stainless steel coved bowls with large radius.
- Two compartment
- Sink can be mounted on top or bottom of countertop
1¹/₂" diameter basket drain in center of bowl. Features³/₄" hole for DI faucet, 1³/₈" hole for mixing faucet and 1¹/₄" hole for eye/drench hose.
Frequently Bought Together