Lab Sinks, Self Rimming, One Bowl, V-3 Series, Eagle MHC
Heavy gauge type 304 series stainless steel coved bowls with large radius.
- One compartment
- Features ³/₄" hole for DI faucet
- 1³/₈" hole for mixing faucet
- 1¹/₄" hole for eye/drench hose
1¹⁄₂"diameter basket drain in center of bowl. Sink can be mounted on top or bottom of countertop.
: SRU sink (10033-360) features an upturn on sides and rear. Top-mount only.
Frequently Bought Together