Lab Sinks, Self Rimming, Two Bowl, V-2 Series, Eagle MHC
Supplier: Eagle Group
Certificates
About this item
Heavy gauge type 304 series stainless steel coved bowls with large radius.
- Sink can be mounted on top or bottom of countertop
- Two compartment
Basket drain of 1½" diameter in center of bowl. Features 1⅜" hole for mixing faucet and 1¼" hole for eye/drench hose.
Frequently Bought Together