Lab Sinks, Self Rimming, One Bowl, V-2 Series, Eagle MHC
Supplier: Eagle Group
Certificates
About this item
Heavy gauge type 304 series stainless steel coved bowls with large radius.
- Sink can be mounted on top or bottom of countertop
One compartment. ³/₂"diameter basket drain in center of bowl. Features ¹¹/₈" hole for mixing faucet and ⁵/₄" hole for eye/drench hose.
Ordering information: SRU sink (10033-226) features an upturn on sides and rear. Top-mount only.
Frequently Bought Together