GeneMate GyroBlotters, Benchmark Scientific
Fournisseur: Ward's
Certificats
À propos de cet article
These platform rockers provide the perfect 3-dimensional motion for all blot related applications. Their tilt angle and speed have been optimally set for gentle, but thorough mixing in gel trays, boxes and other flat vessels.
- 2-year full replacement warranty
- Optimized speed & tilt for gel blotting, washing, and staining
- Tri-directional motion for thorough, gentle mixing
- Incubator & cold room safe
- Flat Rubber Mat Work Surface
The GyroBlotter and GyroBlotter XL can accept stackable platforms and handle loads up to 2 kg. Both have maintenance free brushless motors and are safe for use in cold rooms and incubators.
The GyroBlotter and GyroBlotter XL come standard with autoclavable flat mats and are ideal for blot washes and gel staining/destaining. Both models have been designed to conserve bench space. The GyroBlotter is supplied with a 10.5" x 7.5" platform while the GyroBlotter XL's 12" x 12" platform expands the useable work surface by 80%.
Frequently Bought Together