À propos de cet article
The new Incu-Mixer™ MP series is designed to provide an optimal method for vortexing and incubating microplates. Simultaneous heating from both the bottom surface and the lid provide temperature uniformity throughout the sample. Additionally, the heated lid minimizes evaporation and prevents condensation.
- Optimized for incubated mixing in microplates
- Heated lid prevents condensation
- Accepts plates up to 40 mm high
- Independently control time, temperature and speed
- Available in two-plate or four-plate format
A compact, 2 mm orbit ensures thorough mixing in microplates without splashing. Speed, time and temperature are all displayed in real time on the large LCD screen while an internal validation system constantly monitors all parameters for accuracy.
Designed for application flexibility, the Incu-Mixer™ series is available in two, and four-plate capacities, both capable of accepting all standard plates up to 40 mm high. Independent time, temperature and speed controls allow the mixer to function as three machines in one, a multi-plate incubator, vortexer, or a combination of the two. The Incu-Mixer™ MP series is ideal for a wide variety of molecular biology applications including immuno-chemical reactions, enzyme and protein analysis, and microarray analysis.
Spécifications
- Catalog No:
- CA76323-982
- CA76323-984
- Temperature range:
- Ambient 5…70 °C
- Ambient 5…70 °C
- Temperature accuracy:
- ±0.2 °C
- ±0.2 °C
- Temperature uniformity:
- ±0.5 °C
- ±0.5 °C