Labels, Brady
 :   Avantor
Labels, Brady
 :  Avantor
Labels, Brady
Labels, Brady
Labels for the TLS 2200 and TLS-PC Link Thermal Transfer Portable Printers.
B-488: High Performance Polyester withstands numerous solvents and variable temperatures when applied to various surfaces including Stainless Steel and Polypropylene.
B-461: Self-Laminating Polyester is a clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.
B-499: Nylon Cloth wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.
Please Note: This material is UL, CSA and AGA approved with its respective ribbon. See the 'Recommended Ribbon Series' for the appropriate ribbon to use.
Product Family Options
  • Désignation
    Matière
    Dimensions
    Packaging
  • Portable thermal labels
    Nylon Cloth
    12.7×38.1 mm (0.5×1.5")
    500 labels/roll
    Each
    Each - $103.40
    $103.40
    Each
       $103.40
  • White nylon cloth labels
    Nylon Cloth
    48.26×25.4 mm (1.9×1")
    250 labels/roll
    Each
    Each - $132.30
    $132.30
    Each
       $132.30
  • Portable thermal labels
    High Performance Polyester
    19×22 mm (0.75×0.9")
    250 labels/roll
    Each
    Each - $52.02
    $52.02
    Each
       $52.02
  • Self-laminating labels
    Self-laminating Polyester
    45.72×12.7 mm (1.8×0.5")
    500 labels/roll
    Each
    Each - $117.85
    $117.85
    Each
       $117.85
  • Portable thermal labels
    Self-laminating Polyester
    2.200×0.5" (55.880×12.7 mm)
    100 labels/roll
    Each
    Each - $32.01
    $32.01
    Each
       $32.01