Labels, Brady
 :   Avantor
Labels, Brady
 :  Avantor
Labels, Brady
Labels, Brady
B-499: Nylon Cloth wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable. This material has a permanent acrylic adhesive and is thermal transfer printable.
B-488: High Performance Polyester withstands numerous solvents and variable temperatures when applied to various surfaces including Stainless Steel and Polypropylene.
B-461: Self-Laminating Polyester is a clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.
B-427: Self-Laminating Vinyl is an excellent material for wire and cable identification. This material has good clarity, conformability and is self-extinguishing.
B-351: Tamper-Resistant Vinyl is designed to fracture easily in order to show signs of product tampering and to prevent on-piece label removal.
 : RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved.
Product Family Options
  • Désignation
    Matière
    Pour l'utilisation avec
    Dimensions
  • Printer labels, FNA properties
    Nylon Cloth
    0.5 - 0.7 ml tubes
    19.1 x 12.7 mm
    Each
    Each - $91.15
    $91.15
    Each
       $91.15
  • Printer labels, FNA properties
    Nylon Cloth
    0.5 - 1.5 ml vials
    9.5 mm
    Each
    Each - $68.02
    $68.02
    Each
       $68.02
  • Printer labels, FNA properties
    Nylon Cloth
    2 - 50 ml vials
    12.7 mm
    Each
    Each - $66.70
    $66.70
    Each
       $66.70
  • Printer labels, FNA properties
    Nylon Cloth
    0.5 - 2 ml tube
    22.9 x 12.7 mm
    Each
    Each - $94.50
    $94.50
    Each
       $94.50
  • Printer labels, FNA properties
    Nylon Cloth
    0.5 - 1.5 ml vials
    25.4 x 9.5 x 9.5 mm
    Each
    Each - $139.94
    $139.94
    Each
       $139.94
  • Printer labels, FA properties
    Polyester
    50 ml conical tubes
    48.3 x 25.4 mm
    Each
    Each - $100.06
    $100.06
    Each
       $100.06
  • Printer labels, FA properties
    Polyester
    Well plates
    48.3 x 6.4 mm
    Each
    Each - $132.30
    $132.30
    Each
       $132.30
  • Printer labels, FA properties
    Polyester
    Slides
    22.9 x 22.9 mm
    Each
    Each - $58.97
    $58.97
    Each
       $58.97
  • Printer labels, FNAHSL properties
    Polyester
    0.2 ml tubes
    38.1 x 4.8 x 17.8 mm
    Each
    Each - $117.85
    $117.85
    Each
       $117.85
  • Printer labels, FNAHSL properties
    Polyester
    0.5 - 0.7 ml tubes
    45.7 x 9.5 x 19 mm
    Each
    Each - $89.56
    $89.56
    Each
       $89.56
  • Printer labels, FNAHSL properties
    Polyester
    1 - 8 ml vials
    66.7 x 25.4 x 25.4 mm
    Each
    Each - $55.25
    $55.25
    Each
       $55.25
  • Printer labels, FNAhl properties
    Polyester
    1 - 2 ml tubes
    41.3 x 15.2 x 25.4 mm
    Each
    Each - $68.92
    $68.92
    Each
       $68.92
  • Printer labels, FNAHSL properties
    Polyester
    15 ml conical tubes
    88.9 x 38.1 x 31.8 mm
    Each
    Each - $78.93
    $78.93
    Each
       $78.93
  • Printer labels, FNAHSL properties
    Polyester
    1 - 8 ml vials
    66.7 x 15.2 x 25.4 mm
    Each
    Each - $46.91
    $46.91
    Each
       $46.91
  • Printer labels, FNS properties
    Vinyl
    0.25 ml straw
    25.4 x 25.4 x 9.5 mm
    Each
    Each - $69.15
    $69.15
    Each
       $69.15
  • Printer labels, FNAHSL properties
    Vinyl
    1 - 8 ml vials
    66.7 x 25.4 x 25.4 mm
    Each
    Each - $55.25
    $55.25
    Each
       $55.25
  • Printer labels, F properties
    Vinyl
    Tamper resistant indicating
    38.1 x 25.4 mm
    Each
    Each - $88.94
    $88.94
    Each
       $88.94