Dynalon Polypropylene Beakers, Griffin Style
30620-994
: Dynalon
: 222075-0100
High-quality beakers are made from polypropylene for all routine laboratory requirements. Graduations molded in milliliters.
- Beakers are autoclavable to 121°C
- Special no-drip pouring spout
- Shatter resistant
Chemically resistant to most acids, bases and many common solvents.
Frequently Bought Together