Cryovial® with Lip Seal, Silicon Washer Seal and External Thread, Simport
Supplier: Simport Scientific
Certificates
About this item
Cryovial is designed for storage as low as –196 °C, vapor phase only.
- Sterilized by gamma radiation
- Unique tamper proof
- White marking area for sample identification
- Can be centrifuged up to 17000 ×g
Super fast thread design allowing removal with only 1⁻¹/₄ turn, and an inside thread design that will not contribute to possible contamination. Exclusive silicone washer fitted inside the cap to ensure a positive seal even at the lowest of cryogenic temperatures.
Packaging: Resealable safety-lock bags of 100.
Frequently Bought Together