MultiMax Microcentrifuge Tubes 0.65, 1.7 and 2.0 ml, Sorenson BioScience
Supplier: Sorenson Bioscience
Certificates
About this item
MultiMax microcentrifuge tubes incorporate superior clarity with phenomenal strength. Glass-like clarity allows easy viewing of sample and pellets.
- Temperature rating from –80 to 121 °C
- Positive seal design allows for repeated cap closures and eliminates pressure-induced popping of caps
- Fits all standard rotors
- Autoclavable
Ideal for high-speed centrifugation, proprietary homopolymer polypropylene tubes are rated up to 30000×g. The positive seal design allows repeated cap closures and will not pop during boiling. All tubes feature external graduation marks and frosted writing surfaces. Available in five different colors and rainbow packs.
Frequently Bought Together