Take advantage and trade in ANY brand pipette for a NEW Eppendorf® Research® plus single-channel variable-volume Pipette at a reduced price. Trust Eppendorf's premium products for quality in your laboratory, backed by 75 years of liquid handling experience and green, cutting-edge manufacturing.
- Volume adjustment feature allows desired volume to be reached in few quick turns
- Control button and ejectors low operating force are positioned for ergonomics
- 4-digit, magnifying-shape volume display
- Calibration display and adjustment opening for specific liquids and volumes
- Uses renowned PhysioCare Concept™ thus reduces strain on hand and arm during pipetting, maintains balance, minimal user exertion
- Quick lock makes lower part particularly easy to remove and service (not available for 5 and 10 ml pipettes)
- Spring loaded tip cone promotes tight fit to tip (not available for 5 and 10 ml pipettes)
- Has ability to be autoclaved without disassembling
Repeater Plus pipette has up to 100 repetitive dispensings. Selection of 112 different volumes from 1 L to 10 ml. 20 volume selections per tip.
These reference pipettes have a volume-locking button, no accidental changes. It has separate button for aspiration, dispensing, and tip ejection.
Application: Pipetting of aqueous liquids, two-button operation, mixing liquids, filling of plates, gels, and reaction vessels, phase extraction and removal of supernatants.