Inert Mobile Phase Bottle Cap, Microsolv
Bottle cap, medical grade PTFE insert with 3 holes, PE, Blue, GL-45
Catalog # 97057-126
Supplier: MicroSolv Technology Corp
Supplier #: 58820-06
Jump to:
Product Details & Documents
Most labs use this bottle cap to ensure that their mobile phase is not contaminated and thereby helps to comply with their HPLC GLP requirements
Your mobile phase will only contact medical grade PTFE so there will not be any contamination from plastic or wax film. Keeping these caps on the bottles with the tubing through holes prevents dust and dirt from getting in and minimizes outgassing or absorption of CO2 from the atmosphere. The cap assembly can be autoclaved.
Specifications
- Color:Blue
- Désignation:Bottle cap, medical grade PTFE insert with 3 holes
- Matière:PE
- Epaisseur:GL-45