Trays, Round, Polypropylene, Dynalon
13450-770
: Dynalon
- Cat. no.:13450-770
- Interior Dimensions:10"W x 3"H
- Supplier No.:109844
Fabricated trays of Polypropylene are heavy wall (3/16"), corrosion resistant and may be autoclaved
Trays can be customized with spigots and covers which are fabricated to order
: Additional sizes and materials are available upon request.