Ladle, High Density Polyethylene, Dynalon
: Dynalon
Manufactured from chemically resistant HDPE
Securely welded handle attached to an extra heavy duty bowl with 2 spouts. Handy for sampling and dispensing corrosive liquids.
: Additional sizes are available upon request.
Frequently Bought Together