Calibration Standard, Ion Chromatography, ERA
Catalog # CA11020-070
Supplier: ERA
CAS Number:
Specifications
- Description:Ion Chromatography
- Size:15 ml
- Bromide:0.2-20 mg/L
- Fluoride:0.1-10 mg/L
- Chloride:0.2-20 mg/L
- Nitrate:0.2-20 mg/L
- Phosphate:0.5-30 mg/L
- Sulphate:0.5-30 mg/L
- Cat. no.:CA11020-070
- Supplier No.:981
Specifications
About this item
Use this calibration standard for initial as well as continuing calibration and tuning verification
One 15 mL screw-cap vial yields up to 200 mL of this ion chromatography, calibration standard after dilution. This standard is designed to calibrate or verify IC calibrations.
This standard is provided as a convenient concentrate with densities allowing you to easily perform gravimetric dilutions.
Ordering information: This standard comes with a certificate of traceability and uncertainty.