VWR® Dissection Needle
125 mm (5")
Catalog # 89259-970
Supplier: VWR International
Jump to:
Product Details & Documents
This VWR® Dissection Needle is stainless steel and spear-shaped.
- Stainless steel fabrication
- Spear-shaped needles
These dissection needles have a double-cutting edge.
Specifications
- Longueur:125 mm (5")