VWR® Dissection Set
8-Piece Dissection Set
89259-964
VWR® Dissection Set includes one scalpel knife, one straight-tipped forceps, one curved-tip forceps, one straight teasing needle, one curved teasing needle, one dropper pipet, one dissecting scissor and one ruler.
- Includes a variety of essential dissection tools
Tools in this set are made of durable stainless steel and can be purposed to a variety of different applications.
- Désignation:8-Piece Dissection Set