VWR® Zirconium Crucibles
Fournisseur: VWR International
Certificats
À propos de cet article
VWR® Zirconium Crucibles are low form and are great for high-temperature fusion.
- Heat-resistant up to 900°C
- Available in a range of capacities
- Low-profile design
Zirconium construction is highly resilient against extreme temperatures.
Frequently Bought Together