ContactUsLinkComponent
Soil Standard, Total Petroleum Hydrocarbons (TPH) in Soil, ERA (Waters)
Soil Standard, Total Petroleum Hydrocarbons (TPH) in Soil, ERA (Waters)
Catalog # CA11022-880
Supplier:  ERA
CAS Number:  
Soil Standard, Total Petroleum Hydrocarbons (TPH) in Soil, ERA (Waters)
Catalog # CA11022-880
Supplier:  ERA
Supplier Number:  571
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Certified Reference Material
  • Size:
    50 g
  • Non-polar extractable material:
    300-3,000 mg/kg (TPH) (Gravimetric); 300-3,000 mg/kg (TPH) (IR)
  • Cat. no.:
    CA11022-880
  • Supplier No.:
    571
  • Parameters:
    TPH in Soil
Specifications
About this item
One 50 g screw-cap bottle of soil is ready to be analyzed for total petroleum hydrocarbons (TPH) in soil in the prescence of interfering fatty acids
Use with EPA IR or gravimetric methods 8440 and 9071B.