E-A-Rsoft™ Metal Detectable Corded Earplugs, 3M™
Catalogue # CA89006-808
Fournisseur: 3M
Spécifications
- NRR:32 dB
- Color:Yellow
- Description:Metal detectable corded earplugs
- Cat. no.:CA89006-808
- Packaging:Poly bag
- Supplier No.:311-4106
Spécifications
À propos de cet article
3M™ E-A-Rsoft™ metal detectable earplugs were specially developed for food processing and other industries where it is critical to keep earplugs from contaminating products and processes.
Recognized by most metal-sensing systems, these earplugs feature an implanted stainless steel ball bearing in the plug and metal particles in the cord.