SP Bel-Art Bulb Pipettor, Bel-Art Products, a part of SP
Catalog # 70690-152
Supplier: Bel-Art Products, a Part of SP
Specifications
- Description:Bulb Pipettor
- Cat. no.:70690-152
- Supplier No.:378880000
Specifications
About this item
A tapered, silicone rubber seal provides an airtight fit on all size pipettes.
- Tapered design
- Silicone rubber seal
- Autoclavable
An excellent aspirator for use with Westergren or Wintrobe sedimentation tubes. The bulb can be opened, cleaned, and steam autoclaved at 121 °C (250 °F).
Delivery: Supplied complete with an elastic cord for dedicating the pipettor to a specific reagent bottle.