EVA PAPRs (Powered Air Purifying Respirators), Complete Systems, Bullard®
Supplier: Bullard
Certificates
About this item
The EVA is simple, safe, and dependable.
- Versatile respirator for particulate or chemical environments
- Lightweight, compact, well-balanced design
- Compatible with range of head tops
- Standard comfort belt
In addition, the EVA offers Active Flow Technology™, which maintains an airflow of 8.5 cfm (high speed), 1000 Assigned Protection Factor (depending upon head top), and dual alarms for low flow and low battery. Finally, the EVA has a visual battery fuel gauge to check remaining charge at the push of a button, a brushless motor with ten times the life of a brush motor PAPR, and optional genius charger for battery trending analysis.
The system features one-button operation, a choice of two air speeds, and a single filter design.
Certifications: NIOSH certified.
Ordering information: Complete systems include blower, battery, belt, charger, headtop, breathing tube, and air flow indicator. Filters/cartridges must be ordered separately.
Specifications
- Catalog No:
- 89097-718
- 89097-710
- 89097-706
- 89097-702
- 89097-722
- Airflow:
- 8.5/7.0 cfm (Hi/Lo Speed)
- 8.5/7.0 cfm (Hi/Lo Speed)
- 8.5/7.0 cfm (Hi/Lo Speed)
- 8.5/7.0 cfm (Hi/Lo Speed)
- 8.5/7.0 cfm (Hi/Lo Speed)
- Alarm:
- 77dBA Low Voltage and 77dBA Low Flow
- 77dBA Low Voltage and 77dBA Low Flow
- 77dBA Low Voltage and 77dBA Low Flow
- 77dBA Low Voltage and 77dBA Low Flow
- 77dBA Low Voltage and 77dBA Low Flow
- Battery life:
- 4 - 10 h
- 4 - 10 h
- 4 - 10 h
- 4 - 10 h
- 4 - 10 h
- Battery type:
- Lithium Ion Polymer
- Lithium Ion Polymer
- Lithium Ion Polymer
- Lithium Ion Polymer
- Lithium Ion Polymer
- Motor:
- Brushless
- Brushless
- Brushless
- Brushless
- Brushless