Cutters, Carbide Insert, Excelta
Supplier:   Excelta
Cutters, Carbide Insert, Excelta
Supplier:  Excelta
Certificates
Cutters, Carbide Insert, Excelta
Cutters, Carbide Insert, Excelta
9 Options Available Below
Product Details & Documents
This compound action cutter is ideal for cutting small hard wire.
  • Carbide insert cutting edges for tough hard wire
  • Compound-Action means less pressure on hands
  • Dual leaf springs for ease of use
  • Stainless steel shank for corrosion resistants
These multi-purpose medium round hard wire cutters feature stainless steel construction for corrosion resistance and carbide inserts for maximum blade life. Hard wire cutting capacity is 0.010 to 0.020". The grips are soft cushioned foam and have a surface resistivity of 10¹⁰ ohms/sq. Interestingly, the 149AT is a compound-action cutter, which requires much less force to operate.
Product Family Options
Product Information
  • Désignation
    Matière
    Overall Length
    Curvature
    Availability
    Price
  • Cutters, hard wire, taper head, 2 star quality, carbide inserts, compound action
    Stainless steel
    152 mm (6")
    Log in for availability
    Log in for availability
    Each
    Each - $594.37
    $594.37
    Each
       $594.37
    Supplier Number: 147AT
  • Cutters, taper head, laser flush cut, carbide inserts, short handle
    Carbon steel
    112.5 mm (4.5")
    14.25°
    Log in for availability
    Log in for availability
    Each
    Each - $464.78
    $464.78
    Each
       $464.78
    Supplier Number: C7145X
  • Cutters, taper head, laser flush cut, carbide inserts, molded handle
    Carbon steel
    118.75 mm (4.75")
    14.25°
    Log in for availability
    Log in for availability
    Each
    Each - $437.06
    $437.06
    Each
       $437.06
    Supplier Number: C7245X-GM
  • Cutters, taper head, laser flush cut, carbide inserts, long handle
    Stainless steel
    143.75 mm (5.75")
    14.25°
    Discontinued
    Supplier Number: -
  • Cutters, taper head, laser flush cut, carbide inserts, long handle
    Carbon steel
    143.75 mm (5.75")
    14.25°
    Log in for availability
    Log in for availability
    Each
    Each - $427.76
    $427.76
    Each
       $427.76
    Supplier Number: C7245X
  • Cutters, taper head, laser flush cut, carbide insert, short handle
    Stainless steel
    112.5 mm (4.5")
    14.25°
    Discontinued
    Supplier Number: -
  • Cutters, small round head, hard wire, carbide inserts, cleanroom safe
    Stainless steel
    112.5 mm (4.5")
    12°
    Log in for availability
    Log in for availability
    Each
    Each - $154.72
    $154.72
    Each
       $154.72
    Supplier Number: 2899EHT-CR
  • Cutters, medium round head, hard wire, 2 star quality, carbide inserts
    Stainless steel
    112.5 mm (4.5")
    12°
    Log in for availability
    Log in for availability
    Each
    Each - $199.56
    $199.56
    Each
       $199.56
    Supplier Number: 2897EHT
  • Cutters, medium round head, hard wire, 2 star quality, carbide inserts
    Stainless steel
    112.5 mm (4.5")
    12°
    Log in for availability
    Log in for availability
    Each
    Each - $180.72
    $180.72
    Each
       $180.72
    Supplier Number: 2899EHT
  • Cutters, large round head, hard wire, 2 star quality, carbide inserts
    Stainless steel
    137.5 mm (5.5")
    12°
    Log in for availability
    Log in for availability
    Each
    Each - $199.56
    $199.56
    Each
       $199.56
    Supplier Number: 2896EHT
  • Cutters, small taper head, hard wire, 2 star quality, carbide inserts
    Stainless steel
    106.25 mm (4.25")
    12°
    Log in for availability
    Log in for availability
    Each
    Each - $203.23
    $203.23
    Each
       $203.23
    Supplier Number: 2876EHT