VWR® Bulk Dispensing
Cap and shoe cover bulk dispenser, 12.375×4.5×20.625"
Catalog # 10031-856
Supplier: VWR International
Jump to:
Product Details & Documents
Extra-large capacity maximizes space and reduces restocking time.
- Maximizes space
- Acrylic construction
Acrylic construction with keyhole slots for wall mounting. Features easy top-loading and convenient front-dispensing.
Spécifications
- Désignation:Cap and shoe cover bulk dispenser
- W×D×H:12.375×4.5×20.625"
- Cat. No.:10031-856
- Dispenser type:Wall-mountable
- Material:Acrylic
- No. of compartments:1
- PPE type:Shoe covers|Bouffant caps