VWR® Workstation Organization
VWR® Workstation Organization
Supplier:   VWR International
VWR® Workstation Organization
Supplier:  VWR International
Certificates
VWR® Workstation Organization
VWR® Workstation Organization
2 Options Available Below
Product Details & Documents
Utilize vertical space of your benchtop with the Workstation Riser.
  • Chemically resistant polycarbonate construction eliminates spill worries
  • Clear fabrication provides strength and visibility
  • Easy organization of contents
Put all of your most important tools or frequently used products on the top shelf.
Manufactured with clear 0.5" acrylic (Benchtop Risers) for strength and visibility.
With the Workstation Riser products, take organization to the next level.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Dimensions
    Availability
    Price
  • Benchtop Riser 16" Wide
    Acrylic
    16 x 10 x 10"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $421.90
    $421.90
    Prix promotionnel pour les nouveaux clients
    Each
       $421.90
  • Benchtop Riser 32" Wide
    Acrylic
    32 x 10 x 10"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $732.95
    $732.95
    Prix promotionnel pour les nouveaux clients
    Each
       $732.95
  • Riser Bin
    Polycarbonate
    Discontinued