VWR® Wash Bottle Holders
Supplier: VWR International
Jump to:
À propos de cet article
Maximize bench space with the Quad Bottle Organizer.
- Inert
- Sturdy ABS plastic construction
- Space-saving design
Holds up to four wash bottles. Made of ABS for easy disinfecting with bleach solution.
2 Options Available Below
Product Details & Documents
Maximize bench space with the Quad Bottle Organizer.
- Inert
- Sturdy ABS plastic construction
- Space-saving design
Holds up to four wash bottles. Made of ABS for easy disinfecting with bleach solution.