C-Chip Disposable Hemacytometer, Electron Microscopy Sciences | Avantor
C-Chip Disposable Hemacytometer, Electron Microscopy Sciences
C-Chip Disposable Hemacytometer, Electron Microscopy Sciences
Certificates
C-Chip Disposable Hemacytometer, Electron Microscopy Sciences
C-Chip Disposable Hemacytometer, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The world's first truly disposable Hemacytometer, the C-Chip offers precise volume control and is easy to use.
  • Quartz grade optical plastic
  • Precise design leads to highly reproducible results
  • Two or four counting chambers fitted with clearest grid pattern rich in contrast
  • Exposure to infectious material is reduced due to its closed system
  • Continuous sample containment
  • Light and unbreakable compared to glass
  • No need for cover slips
  • Time and cost saving
  • For single use
Ues for:
Blood Analysis: Blood Cell Counting
Cell Culture: Cell Concentration measurement / Cell Viability Test
Microbiology: Bacteria and Fungal Spore Counting
IVF, IUI: Sperm Counting
Product Family Options
Informations sur le produit
  • Désignation
    Épaisseur
    Chamber Depth
    Taille
    W×L
    Availability
    Price
  • C-Chip Disposable Hemacytometer Fuchs Rosenthal
    1.6 mm
    100, 200 or 10 Micron (Depends on model)
    50 Pack
    25 × 75 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $458.66
    $458.66
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $349.65
    $349.65
    Each
       $458.66
    Pack of 50
       $349.65
    Supplier Number: 63508-03
  • C-Chip Disposable Hemacytometer Burker Turk
    1.6 mm
    100, 200 or 10 Micron (Depends on model)
    50 Pack
    25 × 75 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $463.08
    $463.08
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $353.02
    $353.02
    Each
       $463.08
    Pack of 50
       $353.02
    Supplier Number: 63508-07