ContactUsLinkComponent
Omniprobe 4 Post, Electron Microscopy Sciences
  102773-590
Omniprobe 4 Post, Electron Microscopy Sciences
  102773-590
 :  75964-10
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Ø:
    3 mm
  • Description:
    4 Post
  • Thickness:
    30 µm
  • Cat. no.:
    102773-590
  • Supplier No.:
    75964-10
The 4 post has copper lift-out grids specifically designed for in-situ lift-out. These grids include multiple indexed mounting locations, two with vertical bar attachment surfaces and two with 'V'shaped alighment surfaces. Sides have lower profile for easier access to outermost posts.
 : Packaged in plastic vials.