Omniprobe 3 Post, Electron Microscopy Sciences
Catalog # 102773-586
Supplier: Electron Microscopy Sciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:3 Post Copper Liftout Grids
- Environmentally Preferable:
- Cat. no.:102773-586
- Supplier No.:75964-08
Specifications
About this item
The 3 post copper lift-out grids are specifically designed for in-situ lift-out. These grids include multiple indexed mounting locations with both vertical bar and 'V' shaped attachment surfaces. These grids have a shallower downset and slightly wider center post.
Packaging: Packaged in glass vials.