Fixed Handle 55 Slide Rack, Electron Microscopy Sciences
A stainless steel rack with a fixed handle and grooves for 76×26 mm microscope slides held vertically for maximum use of space and solutions. Accommodates two vertical racks (#71421-VR), which totals 96 slides.
- Désignation:Vertical 55-Slide Rack only