ContactUsLinkComponent
Blumenthal Bone Rongeur, Electron Microscopy Sciences
Catalogue # 100494-558
Minority or Woman-Owned Business Enterprise
undefined
Blumenthal Bone Rongeur, Electron Microscopy Sciences
Catalogue # 100494-558
Supplier Number:  77984-02
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Blumenthal Bone Rongeur, Full Curve
  • Length:
    6"
  • Cat. no.:
    100494-558
  • Supplier No.:
    77984-02
Spécifications
À propos de cet article
Bone Rongeur. Made from stainless steel with a polished finish.
Jaw width: 3 mm
Length: 6"