Omniprobe Xtreme Access Short-Cut™ Probe Tips, Electron Microscopy Sciences | Avantor
Omniprobe Xtreme Access Short-Cut™ Probe Tips, Electron Microscopy Sciences
XA Short Cut Tungsten Probe Tip
Catalog # 102681-284
Fournisseur #:  75960-06
Omniprobe Xtreme Access Short-Cut™ Probe Tips, Electron Microscopy Sciences
Catalog # 102681-284
Omniprobe Xtreme Access Short-Cut™ Probe Tips, Electron Microscopy Sciences
Omniprobe Xtreme Access Short-Cut™ Probe Tips, Electron Microscopy Sciences
XA Short Cut Tungsten Probe Tip
Price
$597.01
Log in pour la disponibilité
Each Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Custom tip designed from tungsten. Tip radius is < 0.5 µm with 6° taper angle for maximum lifetime service. Compatible with Short-Cut™ system for direct conversion to TEM grid
Informations sur les accessoires: Xtreme Access Compatible Probe Point Holder is for use with 1/2" tungsten probe tips. Comes with a storage vessel. The AP250 AutoProbe™ tip holder uses the tungsten probe tip. Compatible with the Short-Cut™ for direct conversion to TEM Grid. Comes with a storage vial and handle.
Spécifications
  • Désignation:
    XA Short Cut Tungsten Probe Tip