ContactUsLinkComponent
Multi Pin Holder for 8 Pin Stubs, Electron Microscopy Sciences
Catalogue # 102098-306
undefined
Multi Pin Holder for 8 Pin Stubs, Electron Microscopy Sciences
Catalogue # 102098-306
Supplier Number:  75952-08H
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Spécifications
  • Description:
    Multiple Holder for 8 Pins Hitachi, M4
  • Cat. no.:
    102098-306
  • Supplier No.:
    75952-08H
Spécifications
À propos de cet article
This 50 mm in diameter multi pin holder is designed to save time. It is made from machined aluminum with stainless steel Allen wrench screws.
It accommodates up to eight standard 12.7 mm pin stubs.