Fuchs-Resenthal Count Chamber, Electron Microscopy Sciences
Product Details & Documents
Designed for counting leukocytes, eosinophils and cells in cerebrospinal fluid.
Grid pattern is etched directly onto the glass surface. The chamber is 0.2 mm deep. Chamber set includes: one chamber and 2 octagonal cover glasses.
Specifications
- Désignation:Fuchs Resenthal Counting Chamber (3720)