Cryo Markers, Electron Microscopy Sciences | Avantor
Cryo Markers, Electron Microscopy Sciences
Cryo Markers, Electron Microscopy Sciences
Certificates
Cryo Markers, Electron Microscopy Sciences
Cryo Markers, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Used for permanently marking cloth, cardboard, and plastic.
It will not fade at ultra-low temperatures. Smudge-proof. Package consist of four, extra fine point pens in assorted colors (red, green, blue, and black).
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Cryo markers
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $65.50
    $65.50
    Log in pour la disponibilité
    Pack of 4
    Pack of 4 - $78.87
    $78.87
    Each
       $65.50
    Pack of 4
       $78.87
    Supplier Number: 62050
  • Cryo markers
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 24
    Case of 24 - $248.24
    $248.24
    Log in pour la disponibilité
    Each
    Each - $248.24
    $248.24
    Case of 24
       $248.24
    Each
       $248.24
    Supplier Number: 62051