Staining Dishes, Electron Microscopy Sciences
Staining Dishes, Electron Microscopy Sciences
Staining Dishes, Electron Microscopy Sciences
Staining Dishes, Electron Microscopy Sciences
A variety of inexpensive staining dishes made from molded glass with and without fitted glass and stainless steel lids.
  • A variety of sizes are available
    Lids and dishes are available separately
Ideal for immersing slides in several solutions.
Product Family Options
  • Désignation
    Dimensions
  • Staining dish, fitted glass Lid
    120×120×50 mm
     : 71423-DL
  • Staining dish, fitted glass lid
    105×86×75 mm
    Each
    Each - $75.87
    $75.87
    Each
       $75.87
     : 71424-DL
  • Staining dish only
    120×122×110 mm
    Each
    Each - $218.90
    $218.90
    Each
       $218.90
     : 71424-D
  • Steel lid only
    120×122×110 mm
    Each
    Each - $120.26
    $120.26
    Each
       $120.26
     : 71424-L