Gel Loading Tips, Electron Microscopy Sciences
Gel Loading Tips, Electron Microscopy Sciences
Certificates
Gel Loading Tips, Electron Microscopy Sciences
Gel Loading Tips, Electron Microscopy Sciences
10 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
KENDALL’s low binding tips ensure accuracy and precision by bonding a proprietary polymer to the resin. Precision and accuracy are increasingly important in molecular biology labs. A pipetter can be 99% accurate and precise, but the tip can com-promise performance by retaining or binding a sample.
  • Reduces the surface energy of liquid and repels any substance
  • Not detectable/Non-reactive
  • Extracted only by a fluorinated solvent
  • Autoclaving will not compromise performance
  • Available in convenient hinged racked package
  • Gel Loading Tips features
Low Binding Polymer Technology: A manufacturing process that binds a perfluorooctyl polymer to polypropylene resin–resulting in an exposed trifluoromethyl surface.
Tips and racks are fully autoclavable. Lengths 83 mm (200 µl tips) or 45 mm (10 µl tips) for access to bottom of narrow vessels, 5 µl graduation for volume QuickCheck. Universal fit for most research-grade pipetters.
Different outside dimensions:
0.57 mm Ext.Ø for loading protein gels. A white rack
0.37 mm Ext.Ø for 0.4 mm thick polyacrylamide sequencing gels. A yellow rack
0.17 mm Ext.Ø for 0.2 mm thick sequencing gels. A blue rack
Product Family Options
Product Information
  • Tip Style
    Volume
    Sterile
    Longueur
    Tip Color
    Packed
    Availability
    Price
  • Round, O.D. 0.57 mm
    5 - 200 µl
    8.3 cm (3.27")
    Blanc
    196 Tips/Rack
    Log in for availability
    Log in for availability
    Each
    Each - $132.93
    $132.93
    Each
       $132.93
    Supplier Number: 72447-02
  • Flat, O.D. 0.37 mm
    5 - 200 µl
    8.3 cm (3.27")
    Jaune
    196 Tips/Rack
    Log in for availability
    Log in for availability
    Each
    Each - $193.45
    $193.45
    Each
       $193.45
    Supplier Number: 72448-01
  • Round, O.D. 0.57 mm
    1 - 10 µL
    +
    4.5 cm (1.77")
    Blanc
    196 Tips/Rack
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 200
    Case of 200 - $135.01
    $135.01
    Log in for availability
    Each
    Each - $135.01
    $135.01
    Case of 200
       $135.01
    Each
       $135.01
    Supplier Number: 72450-02
  • Round, O.D. 0.57 mm
    5 - 200 µl
    8.3 cm (3.27")
    Blanc
    Bulk Packed
    Log in for availability
    Log in for availability
    Log in for availability
    Bag of 500
    Bag of 500 - $181.73
    $181.73
    Log in for availability
    Each
    Each - $181.73
    $181.73
    Bag of 500
       $181.73
    Each
       $181.73
    Supplier Number: 72447-01
  • Flat, O.D. 0.17 mm
    1 - 10 µL
    +
    4.5 cm (1.77")
    Bleu
    196 Tips/Rack
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 200
    Case of 200 - $238.00
    $238.00
    Log in for availability
    Each
    Each - $238.00
    $238.00
    Case of 200
       $238.00
    Each
       $238.00
    Supplier Number: 72452-02
  • Flat, O.D. 0.17 mm
    5 - 200 µl
    +
    8.3 cm (3.27")
    Bleu
    196 Tips/Rack
    Log in for availability
    Log in for availability
    Each
    Each - $250.13
    $250.13
    Each
       $250.13
    Supplier Number: 72449-02
  • Flat, O.D. 0.17 mm
    5 - 200 µl
    8.3 cm (3.27")
    Bleu
    196 Tips/Rack
    Log in for availability
    Log in for availability
    Each
    Each - $219.08
    $219.08
    Each
       $219.08
    Supplier Number: 72449-01
  • Flat, O.D. 0.37 mm
    1 - 10 µL
    +
    4.5 cm (1.77")
    Jaune
    196 Tips/Rack
    Discontinued
    Supplier Number: 72451-02
  • Round, O.D. 0.57 mm
    1 - 10 µL
    4.5 cm (1.77")
    Jaune
    196 Tips/Rack
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 200
    Case of 200 - $132.51
    $132.51
    Log in for availability
    Each
    Each - $132.51
    $132.51
    Case of 200
       $132.51
    Each
       $132.51
    Supplier Number: 72450-01
  • Flat, O.D. 0.17 mm
    1 - 10 µL
    4.5 cm (1.77")
    Bleu
    196 Tips/Rack
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 200
    Case of 200 - $212.32
    $212.32
    Log in for availability
    Each
    Each - $212.32
    $212.32
    Case of 200
       $212.32
    Each
       $212.32
    Supplier Number: 72452-01
  • Round, O.D. 0.57 mm
    5 - 200 µl
    +
    8.3 cm (3.27")
    Blanc
    196 Tips/Rack
    Log in for availability
    Log in for availability
    Each
    Each - $133.81
    $133.81
    Each
       $133.81
    Supplier Number: 72447-03