About this item
Kendall filter pipette tips are certified Rnase / Dnase-Free, Human DNA-Free, and PCR inhibitor-free. Their low binding surface reduces DNA and protection retention. Liquid and aerosol barrier prevent pipetter contamination.
- Types of Membranes
- Aerosol Barrier: Prevents aerosol contamination
- Self-sealing Barrier (contains cellulose): Prevents aerosol and liquid contamination
- Cellulose can contaminate samples
- Liquid Barrier: Prevents aerosol and liquid contamination
- Will not contaminate samples
There's a universal fit on most research-grade pipetters. CellPack keeps tips straight for multi-channel pipetters. Made from high-density polyethylene (naturally hydrophobic). Features aerosol, self-sealing and liquid barrier membranes.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.