Microtube Rack, 4-Way, Electron Microscopy Sciences
Microtube Rack, 4-Way, Electron Microscopy Sciences
Certificates
Microtube Rack, 4-Way, Electron Microscopy Sciences
Microtube Rack, 4-Way, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Create as large a rack as you need by linking together multiple racks. Heavyduty polypropylene racks feature a unique system of tabs and slots, that facilitate easy connection and sturdy fit. Autoclavable. Assorted colors or natural. Measurements: 174 x 95 x 52 mm.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • 4-Way Microtube Rack Natural
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $196.04
    $196.04
    Log in pour la disponibilité
    Pack of 5
    Pack of 5 - $177.01
    $177.01
    Each
       $196.04
    Pack of 5
       $177.01
    Supplier Number: 61615-20
  • 4-Way Microtube Rack Assorted
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $190.11
    $190.11
    Log in pour la disponibilité
    Pack of 5
    Pack of 5 - $152.69
    $152.69
    Each
       $190.11
    Pack of 5
       $152.69
    Supplier Number: 61615-00