Mini-Cooling Chambers, Electron Microscopy Sciences | Avantor
Mini-Cooling Chambers, Electron Microscopy Sciences
Mini-Cooling Chambers, Electron Microscopy Sciences
Certificates
Mini-Cooling Chambers, Electron Microscopy Sciences
Mini-Cooling Chambers, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ideal for ice pans and ice buckets for use when only a small number of microtubes are required.
Each Mini-Chamber will fit in our 1L Glacierbrand™ Ice Pan (6.13 x 6.13 x 3.61"). Minimize Ice - Less Ice is needed to cool these chambers. Less ice means less mess. Mini-Chambers are anodized aluminum and utilize our "Surround-A-Tube" cooling technology.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Mini Cooling Chamber, 6 Holes
    Discontinued
    Supplier Number: 68600-10
  • Mini Cooling Chamber , 15 Holes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $721.81
    $721.81
    Each
       $721.81
    Supplier Number: 68600-20
  • Mini Cooling Chamber 30 Holes
    Discontinued
    Supplier Number: 68600-30