Glacier Ice Buckets, Electron Microscopy Sciences | Avantor
Glacier Ice Buckets, Electron Microscopy Sciences
Glacier Ice Buckets, Electron Microscopy Sciences
Certificates
Glacier Ice Buckets, Electron Microscopy Sciences
Glacier Ice Buckets, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Excellent Insulation: Made of the same triple density expanded polystyrene as our popular Ice Pans, preserve ice 30% longer.
  • Won't Sweat - Thick insulating walls will not sweat
  • Easy Grip - Robust handles make buckets easy to carry
  • Includes Lid - Use the supplied lid to maximize cooling
  • Heavy Duty - Solidly constructed to be unbreakable under normal laboratory conditions
  • Chemically Resistant - Resistant to mild chemicals* as well as ice-alcohol slurry, dry ice and liquid nitrogen
(*Not recommended for long-term exposure to harsh chemicals or corrosives.)
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Color
    Hauteur
    Top Diameter
    Availability
    Price
  • Ice bucket, small
    2.5 L
    Blue
    4.375"
    10.625"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $115.81
    $115.81
    Each
       $115.81
    Supplier Number: 68600-40
  • 10.625"
    Discontinued
    Supplier Number: -
  • Ice bucket, large
    5 L
    Blue
    6.250"
    10.625"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $148.94
    $148.94
    Each
       $148.94
    Supplier Number: 68600-60
  • 10.625"
    Discontinued
    Supplier Number: -