Electropolished Stainless Steel Freezer Racks , Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
À propos de cet article
High Quality, Ultra-Durable Freezer Racks for Long Term Sample Storage.
- No Sharp Edges - All edges have been ground down to remove the sharp surfaces that plague standard freezer racks
- Electropolished - This step cleans up all weld points that, over time, may be the starting points for corrosion
- Superior Construction - Superior construction and design have created a more sturdy rack than what is currently available on the market
- Durable - Racks are designed and finished to last longer in the demanding freezer environment where samples are housed
- Surgical Finish - Electropolishing leaves racks ultra-shiny and free from any surface contaminants
- Multiple Sizes - Several vertical and horizontal options are available to fit a variety of freezer configurations
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together