EMS Counting Cells for Haemocytometry, Electron Microscopy Sciences | Avantor
EMS Counting Cells for Haemocytometry, Electron Microscopy Sciences
EMS Counting Cells for Haemocytometry, Electron Microscopy Sciences
Certificates
EMS Counting Cells for Haemocytometry, Electron Microscopy Sciences
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All complete with case and 1 pair of cover glasses.
The exceptions are the Howard Cell which doesn't include a cover glass and the Sedgewick Rafter which includes one cover glass.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Malassez Double Cell 0.2 mm Standard French Rule
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $390.78
    $390.78
    Each
       $390.78
    Supplier Number: 68052-10
  • Thoma Double Cell 0.1 mm Standard German Rule
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $494.95
    $494.95
    Each
       $494.95
    Supplier Number: 68052-12
  • Neubauer Double Cell 0.1 mm, Modified Thoma Rule
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $723.44
    $723.44
    Each
       $723.44
    Supplier Number: 68052-14
  • Improved Neubauer Double Cell 0.1 mm, Ruling allows full use
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $500.78
    $500.78
    Each
       $500.78
    Supplier Number: 68052-16
  • Burker Double Cell 0.1 mm, For counts in bands
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $386.66
    $386.66
    Each
       $386.66
    Supplier Number: 68052-17