EMS Engraver, Electron Microscopy Sciences
Permanently be able to mark all of your important tools and samples.
The Engraver comes with a battery pack and is operated on two "AA" batteries. Our EMS Engraver engraves test tubes, bottles, racks, or keys as well as anything made from glass, metal, stone as well as bone. Engravings are permanent and will not disappear in the presence of chemicals and solvents. The Engraver is 17.8 cm (7") in Height. We offer a stand for the EMS Engraver that is made from polypropylene and it measures (3.5 x 3.375").
Permanently be able to mark all of your important tools and samples.
The Engraver comes with a battery pack and is operated on two "AA" batteries. Our EMS Engraver engraves test tubes, bottles, racks, or keys as well as anything made from glass, metal, stone as well as bone. Engravings are permanent and will not disappear in the presence of chemicals and solvents. The Engraver is 17.8 cm (7") in Height. We offer a stand for the EMS Engraver that is made from polypropylene and it measures (3.5 x 3.375").
Product Family Options
- Désignation
- : 72096-01
- Engraver tool stand-: -