Lighted Forceps, Scissors, Electron Microscopy Sciences | Avantor
Lighted Forceps, Scissors, Electron Microscopy Sciences
Lighted Forceps, Scissors, Electron Microscopy Sciences
Certificates
Lighted Forceps, Scissors, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
See your samples and your embeddings in a whole new light!
Increases the visibility of the specimen
Allows for accurate orientation of tissue and specimen
Minimizers floaters
Reduces embedding and prep time
These lighted forceps ease identification and location of otherwise undetectable tissue fragments, while reducing embedding time. Increases the visibility of specimen allowing for more accurate work. Made of German stainless steel, are submersible and light longevity approx. 20,000 hours.
Delivery information: Each kit includes: one instrument, power pack and battery charger.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Tip Style
    Availability
    Price
  • Forceps
    15 cm
    Smooth Fine Tip
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $676.32
    $676.32
    Log in pour la disponibilité
    Kit of 2
    Kit of 2 - $676.32
    $676.32
    Each
       $676.32
    Kit of 2
       $676.32
    Supplier Number: 77905-02
  • Surgical Scissor
    17 cm
    Blunt/Blunt Tip
    Discontinued
    Supplier Number: 77905-08