StatMark Pen, Electron Microscopy Sciences
About this item
StatMark Pens are made of rigid nylon that allow for precise hand marking on glass slides as well as on most tissue embedding cassettes.
- Permanent ink in an ultrafine tip
- Quick drying ink is highly resistant to aqueous based stains, alcohols, xylene and xylene substitutes
- Do not dry out easily
- Last twice as long as many of the leading 'laboratory' pens
StatMark Pens are made of rigid nylon that allow for precise hand marking on glass slides as well as on most tissue embedding cassettes. This quick drying ink is highly resistant to aqueous based stains, alcohols, xylene and xylene substitutes. StatMark Pens do not dry out easily and last twice as long as many of the leading 'laboratory' pens.
2 Options Available Below
Product Details & Documents
StatMark Pens are made of rigid nylon that allow for precise hand marking on glass slides as well as on most tissue embedding cassettes.
- Permanent ink in an ultrafine tip
- Quick drying ink is highly resistant to aqueous based stains, alcohols, xylene and xylene substitutes
- Do not dry out easily
- Last twice as long as many of the leading 'laboratory' pens
StatMark Pens are made of rigid nylon that allow for precise hand marking on glass slides as well as on most tissue embedding cassettes. This quick drying ink is highly resistant to aqueous based stains, alcohols, xylene and xylene substitutes. StatMark Pens do not dry out easily and last twice as long as many of the leading 'laboratory' pens.
Product Family Options
Product Information
- DésignationAvailabilityPrice
- Supplier Number: 72109-01
- StatMark PenEach$147.15Pack of 12$142.94Supplier Number: 72109-12