Staining Dish Set for 30 Slides, Electron Microscopy Sciences
À propos de cet article
These mix and match staining dish components offer greater flexibility in meeting your requirements. This staining dish accommodates 16, 20 and 30 slide racks.
- Dish is made from soda-lime glass
- Holds a variety of sizes: 3 x 1", 3 x 11/2 ", and 3 x 2"
- Inside: 135 (H) x 102 (W) x 69 (D) mm
- Outisde: 146 (H) x 113 (W) x 83 (D) mm
Delivery information: Set includes of one staining dish, one lid, one 30-slide stainless steel rack.
3 Options Available Below
Product Details & Documents
These mix and match staining dish components offer greater flexibility in meeting your requirements. This staining dish accommodates 16, 20 and 30 slide racks.
- Dish is made from soda-lime glass
- Holds a variety of sizes: 3 x 1", 3 x 11/2 ", and 3 x 2"
- Inside: 135 (H) x 102 (W) x 69 (D) mm
- Outisde: 146 (H) x 113 (W) x 83 (D) mm
Delivery information: Set includes of one staining dish, one lid, one 30-slide stainless steel rack.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: 70312-30
- Supplier Number: 70312-31
- Staining Dish Cover OnlyDiscontinued-Supplier Number: 70312-32
- Supplier Number: 70312-33