Histo-Mark Pen, Electron Microscopy Sciences | Avantor
Histo-Mark Pen, Electron Microscopy Sciences
Histo-Mark Pen, Electron Microscopy Sciences
Certificates
Histo-Mark Pen, Electron Microscopy Sciences
Histo-Mark Pen, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The fine-line Histo-Mark pen is designed to work like a pen.
  • Can be used on any surface
  • Marking quality remains consistent throughout the life of the ink barrel
The Histo-Mark pen is solvent resistent, especially to alcohol, acetone, xylene, xylene subsite and toluene.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Point Type
    Availability
    Price
  • Histo-Mark pen
    Black
    Fine
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $19.58
    $19.58
    Each
       $19.58
    Supplier Number: 72155-01
  • Histo-Mark pen, set of 12
    Black
    Fine
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 12
    Box of 12 - $159.69
    $159.69
    Log in pour la disponibilité
    Each
    Each - $159.69
    $159.69
    Box of 12
       $159.69
    Each
       $159.69
    Supplier Number: 72155-12